Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 2 (d2) 10mg
Supplier:  ATCC 501X
Encompass
Purified pneumococcal polysaccharide type 2 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 2 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2768