Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 5 (d5) 200mg
Catalog No. 502382386
Encompass
Purified pneumococcal polysaccharide type 5 is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 5 (200 mg) is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 5 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2386 Supplier ATCC Supplier No. 41X
Add to Cart
Add to Cart
Purified pneumococcal polysaccharide type 5 is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 5 (200 mg) is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 5 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.