Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 3 (d3) 200mg
Catalog No. 502382740
Encompass
Purified pneumococcal polysaccharide type 3 is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 3 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 3 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2740 Supplier ATCC Supplier No. 33X
Purified pneumococcal polysaccharide type 3 is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 3 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 3 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.