Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 43 (d11A) 10mg
Catalog No. 502382033
Encompass
Purified pneumococcal polysaccharide type 11A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 11A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2033 Supplier ATCC Supplier No. 513X
Purified pneumococcal polysaccharide type 11A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 11A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.