Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 57 (d19A) 2mg
Catalog No. 502382778
Encompass
Purified pneumococcal polysaccharide type 19A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 19A is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 19A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2778 Supplier ATCC Supplier No. 301X
Purified pneumococcal polysaccharide type 19A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 19A is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 19A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.