Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 54 (d15B) 10mg
Catalog No. 502384024
Encompass
Purified pneumococcal polysaccharide type 15B is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 15B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4024 Supplier ATCC Supplier No. 519X
Purified pneumococcal polysaccharide type 15B is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 15B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.