Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 8 (d8) 200mg
Supplier:  ATCC 505X
Encompass
Purified pneumococcal polysaccharide type 8 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 8 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4082