Learn More
ATCC pfizer Pneumococcal polysaccharide powder Type 14 (d14) 2mg
Catalog No. 502384250
Encompass
Purified pneumococcal polysaccharide type 14 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 14 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 14 can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.
Catalog No. 50-238-4250 Supplier ATCC Supplier No. 73X
Purified pneumococcal polysaccharide type 14 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 14 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 14 can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.