Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 20 (d20) 200mg
Supplier:  ATCC 526X
Encompass
Purified pneumococcal polysaccharide type 20 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 20 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4577
 Restricted Item