Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 9 (d9N) 10mg
Supplier:  ATCC 507X
Encompass
Purified pneumococcal polysaccharide type 9N is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 9N can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4595