Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 17 (d17F) 200mg
Supplier:  ATCC 523X
Encompass
Purified pneumococcal polysaccharide type 17F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 17F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4189