Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 26 (d6B) 200mg
Supplier:  ATCC 107X
Encompass
Purified pneumococcal polysaccharide type 6B is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 6B is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 6B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-5107