Learn More
ATCC pfizer Pneumococcal polysaccharide powder Type 2 (d2) 200mg
Catalog No. 502382769
Encompass
Purified pneumococcal polysaccharide type 2 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 2 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2769 Supplier ATCC Supplier No. 502X
Add to Cart
Add to Cart
Purified pneumococcal polysaccharide type 2 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 2 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.