Learn More
ATCC Pfizer Pneumococcal; polysaccharide powder; Type 66 (35B) 10mg
Catalog No. 502380019
Encompass
Purified pneumococcal polysaccharide type 35B is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 35B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-0019 Supplier ATCC Supplier No. 540X
Purified pneumococcal polysaccharide type 35B is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 35B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.