Learn More
ATCC Pfizer Pneumococcal; polysaccharide powder; Type 66 (35B) 2 mg
Catalog No. 502380014
Encompass
Purified pneumococcal polysaccharide type 35B is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 35B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-0014 Supplier ATCC Supplier No. 539X
Purified pneumococcal polysaccharide type 35B is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 35B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.