Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 6 (d6A) 2mg
Catalog No. 502382058
Encompass
Purified pneumococcal polysaccharide type 6A is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 6A is a component of the current 13-valent vaccine against Streptococcus pneumoniae. Pneumococcal polysaccharide type 6A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2058 Supplier ATCC Supplier No. 14X
Purified pneumococcal polysaccharide type 6A is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 6A is a component of the current 13-valent vaccine against Streptococcus pneumoniae. Pneumococcal polysaccharide type 6A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.