Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 5 (d5) 10mg
Catalog No. 502382603
Encompass
Purified pneumococcal polysaccharide type 5 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 5 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 5 can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.
Catalog No. 50-238-2603 Supplier ATCC Supplier No. 38X
Purified pneumococcal polysaccharide type 5 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 5 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 5 can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.