Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 12 (d12F) 10mg
Catalog No. 502383293
Encompass
Purified pneumococcal polysaccharide type 12F is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 12F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-3293 Supplier ATCC Supplier No. 516X
Purified pneumococcal polysaccharide type 12F is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 12F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.