Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 6 (d6A) 10mg
Catalog No. 502383793
Encompass
Purified pneumococcal polysaccharide type 6A (US Type 6) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 6A is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 6A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-3793 Supplier ATCC Supplier No. 271X
Purified pneumococcal polysaccharide type 6A (US Type 6) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 6A is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 6A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.