Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 22 (d22F) 2 mg
Catalog No. 502384849
Encompass
Purified pneumococcal polysaccharide type 22F (US type 22) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 22F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4849 Supplier ATCC Supplier No. 527X
Purified pneumococcal polysaccharide type 22F (US type 22) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 22F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.