Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 70 (d33F) 10mg
Catalog No. 502382604
Encompass
Purified pneumococcal polysaccharide type 33F (US type 70) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 33F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2604 Supplier ATCC Supplier No. 534X
Purified pneumococcal polysaccharide type 33F (US type 70) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 33F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.