Learn More
ATCC pfizer Pneumococcal polysaccharide powder Type 19 (d19F) 10mg
Catalog No. 502383502
Encompass
Purified pneumococcal polysaccharide type 19F is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 19F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 19F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-3502 Supplier ATCC Supplier No. 99X
Purified pneumococcal polysaccharide type 19F is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 19F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 19F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.