Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 68 (d9V) 2mg
Catalog No. 502383635
Encompass
Purified pneumococcal polysaccharide type 9V is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 9V is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 9V can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.
Catalog No. 50-238-3635 Supplier ATCC Supplier No. 305X
Purified pneumococcal polysaccharide type 9V is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 9V is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 9V can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.