Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 23 (d23F) 10mg
Catalog No. 502382171
Encompass
Purified pneumococcal polysaccharide type 23F is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 23F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. Pneumococcal polysaccharide type 23F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2171 Supplier ATCC Supplier No. 103X
 Restricted Item
Add to Cart
Add to Cart
Purified pneumococcal polysaccharide type 23F is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 23F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. Pneumococcal polysaccharide type 23F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.