Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 51 (d7F) 10mg
Catalog No. 502384103
Encompass
Purified pneumococcal polysaccharide type 7F (US Type 51) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 7F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 7F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4103 Supplier ATCC Supplier No. 281X
Purified pneumococcal polysaccharide type 7F (US Type 51) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 7F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 7F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.