Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 20 (d20) 10mg
Supplier:  ATCC 525X
Encompass
Purified pneumococcal polysaccharide type 20 (US type 20) is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal polysaccharide type 20 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-1851