Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 26 (d6B) 10mg
Catalog No. 502382835
Encompass
Purified pneumococcal polysaccharide type 6B is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 6B is a component of the current 13-valent vaccine against Streptococcus pneumoniae. Pneumococcal polysaccharide type 6B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2835 Supplier ATCC Supplier No. 106X
Purified pneumococcal polysaccharide type 6B is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 6B is a component of the current 13-valent vaccine against Streptococcus pneumoniae. Pneumococcal polysaccharide type 6B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.