Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 23 (d23F) 2mg
Supplier:  ATCC 102X
Encompass
Purified pneumococcal polysaccharide type 23F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 23F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 23F can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.
Catalog No. 50-238-4551