Learn More
ATCC Pfizer Pneumococcal; polysaccharide powder; Type 46 (23A) 200 mg
Catalog No. 502379608
Encompass
Purified pneumococcal polysaccharide type 23A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 23A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-237-9608 Supplier ATCC Supplier No. 547X
Purified pneumococcal polysaccharide type 23A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 23A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.