Learn More
ATCC PfizerPneumococcal polysaccharide powder Type 51 (d7F) 2mg
Catalog No. 502384839
Encompass
Purified pneumococcal polysaccharide type 7F is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 7F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 7F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-4839 Supplier ATCC Supplier No. 108X
Purified pneumococcal polysaccharide type 7F is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 7F is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 7F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.