Learn More
TARGETMOL CHEMICALS INC PFK-015 25MG
SDP
Catalog No. 502580800
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PFK-015 (PFK15) is an effective inhibitor of PFKFB3 (IC50 110 nM) and inhibits PFKFB3 activity in Y cells (IC50 20 nM). purity: 99%
Catalog No. 50-258-0800 Supplier TARGETMOL CHEMICALS INC Supplier No. T245525MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PFK-015 (PFK15) is an effective inhibitor of PFKFB3 (IC50 110 nM) and inhibits PFKFB3 activity in Y cells (IC50 20 nM). purity: 99%