Learn More
Apexbio Technology LLC PFK-015 4382-63-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089545
Encompass_Preferred
PFK-015 (CAS 4382-63-2) is a selective inhibitor of 6-phosphofructo-2-kinase/fructose-2 6-biphosphatase 3 (PFKFB3) with an IC50 of 207 nM PFKFB3 regulated by HIF- and activated by oncogenic signaling pathways such as AKT and Ras plays a key role in glycolytic flux and tumor metabolism In cellular assays PFK-015 exhibits cytotoxic effects in lung adenocarcinoma (H522) and leukemia (Jurkat) cell lines suppressing glucose uptake intracellular ATP and F2 6BP levels It promotes apoptosis in Jurkat cells and reduces metastatic spread in a Lewis lung carcinoma mouse model PFK-015 is utilized in cancer research to investigate glycolysis inhibition as a therapeutic strategy
Catalog No. 50-000-89545 Supplier Apexbio Technology LLC Supplier No. B35755.1
PFK-015 (CAS 4382-63-2) is a selective inhibitor of 6-phosphofructo-2-kinase/fructose-2 6-biphosphatase 3 (PFKFB3) with an IC50 of 207 nM PFKFB3 regulated by HIF- and activated by oncogenic signaling pathways such as AKT and Ras plays a key role in glycolytic flux and tumor metabolism In cellular assays PFK-015 exhibits cytotoxic effects in lung adenocarcinoma (H522) and leukemia (Jurkat) cell lines suppressing glucose uptake intracellular ATP and F2 6BP levels It promotes apoptosis in Jurkat cells and reduces metastatic spread in a Lewis lung carcinoma mouse model PFK-015 is utilized in cancer research to investigate glycolysis inhibition as a therapeutic strategy