Learn More
Selleck Chemical LLC PFK158-5mg
SDP
Supplier:  Selleck Chemical LLC S88075MG
Encompass_Preferred
PFK158 is a potent and selective inhibitor of PFKFB3. It has improved PK properties and causes ~80% growth inhibition in several mouse models of human-derived tumors.
Catalog No. 50-238-9912