Learn More
Aviva Systems Biology Corp PFKFB1 Antibody - C-terminal region (ARP61878_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61878_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PFKFB1. It was validated on Western Blot. . Alternate names: F6PK, HL2K, MGC116715, MGC116717, PFRXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-494