Learn More
Sino Biological Recombinant Human PADI1 Protein (Full Length, GST Tag), Active
Catalog No. 502573751
Encompass_Preferred
Recombinant full length human PAD1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3751 Supplier Sino Biological Supplier No. P312310G50
Recombinant full length human PAD1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.