Learn More
Aviva Systems Biology Corp PFKM Antibody - C-terminal region (ARP61167_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61167_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PFKM. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PFKM. Alternate names: GSD7, MGC8699, PFK-1, PFK1, PFKA, PFKXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-280