Learn More
Medchemexpress LLC PFKM Human HEK293 100ug
SDP
Catalog No. 5000196482
Encompass_Preferred
PFKM protein is a recombinant human phosphofructokinase (muscle isoform) produced in HEK293 cells and provided as a filtered, stabilized solution for enzymatic and biochemical studies. The preparation is high purity and characterized for activity, and it is supplied in multiple pack sizes with recommended storage at -80°C to preserve function.
  • Expressed in HEK293 cells with C-terminal His tag.
  • Supplied in buffered formulation with trehalose for stability.
  • High purity suitable for biochemical and enzymatic assays.
  • Validated specific activity for functional assays.
  • Stable when stored at -80°C; avoid repeated freeze-thaw.
Catalog No. 50-001-96482 Supplier Medchemexpress LLC Supplier No. HYP71090100UG
PFKM protein is a recombinant human phosphofructokinase (muscle isoform) produced in HEK293 cells and provided as a filtered, stabilized solution for enzymatic and biochemical studies. The preparation is high purity and characterized for activity, and it is supplied in multiple pack sizes with recommended storage at -80°C to preserve function.
  • Expressed in HEK293 cells with C-terminal His tag.
  • Supplied in buffered formulation with trehalose for stability.
  • High purity suitable for biochemical and enzymatic assays.
  • Validated specific activity for functional assays.
  • Stable when stored at -80°C; avoid repeated freeze-thaw.