Learn More
Medchemexpress LLC PFKM Human HEK293 10ug
SDP
Catalog No. 5000196110
Encompass_Preferred
Recombinant human PFKM protein expressed in HEK293 cells with a C-terminal His tag, supplied as a 0.2 μm filtered solution and suitable for biochemical and functional assays; catalyzes phosphorylation of fructose-6-phosphate in glycolysis.
  • Human source expressed in HEK293 cells for mammalian post-translational modifications.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Supplied in buffered formulation with trehalose for enhanced stability.
  • Stable at -80°C; avoid repeated freeze-thaw cycles.
  • Validated specific activity: 502.29 pmol/min/μg.
Catalog No. 50-001-96110 Supplier Medchemexpress LLC Supplier No. HYP7109010UG
Recombinant human PFKM protein expressed in HEK293 cells with a C-terminal His tag, supplied as a 0.2 μm filtered solution and suitable for biochemical and functional assays; catalyzes phosphorylation of fructose-6-phosphate in glycolysis.
  • Human source expressed in HEK293 cells for mammalian post-translational modifications.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Supplied in buffered formulation with trehalose for enhanced stability.
  • Stable at -80°C; avoid repeated freeze-thaw cycles.
  • Validated specific activity: 502.29 pmol/min/μg.