Learn More
Carbosynth LLC. Mouse monoclonal PfLDH antibody
SDP
Supplier:  Carbosynth LLC. 10P09DS1MG
Encompass_Preferred
Lactate dehydrogenase catalyzes the interconversion of pyruvate and lactate with concomitant interconversion of NADH and NAD+. It converts pyruvate, the final product of glycolysis to lactate when oxygen is absent or in short supply, and it performs the reverse reaction during the Cori cycle in the liver. Plasmodium falciparum is a protozoan parasite, one of the species of Plasmodium that cause malaria in humans. It is transmitted by the female Anopheles mosquito.
Catalog No. 50-125-2657