Learn More
Medchemexpress LLC PfLDH P. falciparum 10ug | 10ug
SDP
Catalog No. 5000185873
Encompass_Preferred
The pfHPRT (Plasmodium falciparum hypoxanthine-guanine phosphoribosyltransferase) protein is a member of the LDH/MDH (lactate dehydrogenase/malate dehydrogenase) superfamily It plays a crucial role in the purine metabolism salvage pathway of the malaria-causing parasite Plasmodium falciparum PfLDH Protein P falciparum (His) is the recombinant PfLDH protein expressed by E coli with C-His labeled tag
Catalog No. 50-001-85873 Supplier Medchemexpress LLC Supplier No. HYP7713410UG
The pfHPRT (Plasmodium falciparum hypoxanthine-guanine phosphoribosyltransferase) protein is a member of the LDH/MDH (lactate dehydrogenase/malate dehydrogenase) superfamily It plays a crucial role in the purine metabolism salvage pathway of the malaria-causing parasite Plasmodium falciparum PfLDH Protein P falciparum (His) is the recombinant PfLDH protein expressed by E coli with C-His labeled tag