Learn More
New England Biolabs, Inc. PflFI – 2000 units
SDP
Catalog No. 50812312
Encompass
A restriction endonuclease that recognizes the sequence GACN^N_NGTC.
Catalog No. 50-812-312 Supplier New England Biolabs, Inc. Supplier No. R0595S
A restriction endonuclease that recognizes the sequence GACN^N_NGTC.