Learn More
New England Biolabs, Inc. PflMI – 1000 units
SDP
Catalog No. 50812156
Encompass
A restriction endonuclease that recognizes the sequence CCAN_NNN^NTGG.
Catalog No. 50-812-156 Supplier New England Biolabs, Inc. Supplier No. R0509S
A restriction endonuclease that recognizes the sequence CCAN_NNN^NTGG.