Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Tubing Connector, PVDF, 1/4 in., Tri-Clamp, 25/Pk.
Catalog No. NC3440470
Encompass_Preferred
The Saint-Gobain Pure-Fit SIB Tubing Connector is a smooth inner bore hose-barb connector designed to provide seamless transitions between tubing and fittings. Constructed from PVDF, it ensures fluid integrity in biopharmaceutical applications through its smooth inner bore design and patented 360° compression seal.
  • Smooth inner bore (SIB®) design eliminates entrapment and leak points.
  • Complete 360° compression provides a secure seal.
  • Eliminates turbulence or stagnation, preventing bacteria growth.
  • Patented retention and seal system ensures reliable performance.
  • Compatible with all flexible tubing.
  • Autoclavable and sterilizable.
  • Manufactured and packaged in a certified clean room.
  • All materials are animal-free and BPA free.
  • Prevents fluid contamination and reduces product loss.
  • Reduces risk of tubing damage.
Catalog No. NC3440470 Supplier Accuflow Systems Supplier No. PFLS250PVDF
The Saint-Gobain Pure-Fit SIB Tubing Connector is a smooth inner bore hose-barb connector designed to provide seamless transitions between tubing and fittings. Constructed from PVDF, it ensures fluid integrity in biopharmaceutical applications through its smooth inner bore design and patented 360° compression seal.
  • Smooth inner bore (SIB®) design eliminates entrapment and leak points.
  • Complete 360° compression provides a secure seal.
  • Eliminates turbulence or stagnation, preventing bacteria growth.
  • Patented retention and seal system ensures reliable performance.
  • Compatible with all flexible tubing.
  • Autoclavable and sterilizable.
  • Manufactured and packaged in a certified clean room.
  • All materials are animal-free and BPA free.
  • Prevents fluid contamination and reduces product loss.
  • Reduces risk of tubing damage.