Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Tri-Clamp Hose-Barb Connector, PVDF, 3/8 in
Catalog No. NC3440473
Encompass_Preferred
The Saint-Gobain Pure-Fit SIB Tri-Clamp connector is a smooth inner bore (SIB) hose-barb fitting designed to provide a seamless transition between tubing and Tri-Clamp fittings. The SIB geometry minimizes dead volume and potential fluid stagnation, improving flow profiles and reducing sites for contamination.
  • Smooth inner bore reduces dead volume and turbulence.
  • PVDF construction provides chemical resistance and low extractables.
  • 3/8 in (9.5 mm) hose-barb for secure tubing connection.
  • Tri-Clamp connection for sanitary system integration.
  • Manufactured and packaged in a certified clean room.
Catalog No. NC3440473 Supplier Accuflow Systems Supplier No. PFLS375PVDF
The Saint-Gobain Pure-Fit SIB Tri-Clamp connector is a smooth inner bore (SIB) hose-barb fitting designed to provide a seamless transition between tubing and Tri-Clamp fittings. The SIB geometry minimizes dead volume and potential fluid stagnation, improving flow profiles and reducing sites for contamination.
  • Smooth inner bore reduces dead volume and turbulence.
  • PVDF construction provides chemical resistance and low extractables.
  • 3/8 in (9.5 mm) hose-barb for secure tubing connection.
  • Tri-Clamp connection for sanitary system integration.
  • Manufactured and packaged in a certified clean room.