Learn More
Medchemexpress LLC HY-116770 5mg Medchemexpress, PFM01 CAS:1558598-41-6 Purity:>98%
SDP
Catalog No. 502080895
Encompass_Preferred
Medchemexpress, HY-116770 5mg PFM01 CAS:1558598-41-6 PFM01, N-alkylated Mirin derivative, is a MRE11 endonuclease inhibitor. PFM01 can regulate double-strand break repair (DSBR) by nonhomologous end-joining (NHEJ) versus homologous recombination (HR). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0895 Supplier Medchemexpress LLC Supplier No. HY1167705MG
Medchemexpress, HY-116770 5mg PFM01 CAS:1558598-41-6 PFM01, N-alkylated Mirin derivative, is a MRE11 endonuclease inhibitor. PFM01 can regulate double-strand break repair (DSBR) by nonhomologous end-joining (NHEJ) versus homologous recombination (HR). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.