Learn More
Medchemexpress LLC PFM39 | 1310744-67-2 | 99.0% | 50 MG
SDP
Catalog No. 5000232090
Encompass_Preferred
PFM39 is a Mirin analog that selectively inhibits the exonuclease activity of MRE11 and is supplied for research use only.
  • Selective MRE11 exonuclease inhibitor suitable for biochemical assays.
  • High purity (99.0%) for reliable experimental results.
  • Molecular weight 219.26 g/mol and formula C10H9N3OS.
  • Available as purified solid or 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-32090 Supplier Medchemexpress LLC Supplier No. HY12095150MG
Add to Cart
Add to Cart
PFM39 is a Mirin analog that selectively inhibits the exonuclease activity of MRE11 and is supplied for research use only.
  • Selective MRE11 exonuclease inhibitor suitable for biochemical assays.
  • High purity (99.0%) for reliable experimental results.
  • Molecular weight 219.26 g/mol and formula C10H9N3OS.
  • Available as purified solid or 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.