Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Connector, PVDF, Mini, 1/2 in
Catalog No. NC3440482
Encompass_Preferred
The Pure-Fit SIB Mini PVDF smooth inner-bore connector provides a seamless tubing-to-fitting transition for aseptic fluid transfer in bioprocessing and laboratory systems. Constructed from PVDF, the Mini design minimizes dead volume and reduces particle entrapment, supporting low extractables and biocompatibility.
  • Smooth inner bore reduces dead volume and particle entrapment.
  • PVDF construction for broad chemical compatibility and sterilization tolerance.
  • Mini series, 1/2 in (12.7 mm) size for common tubing applications.
  • Manufactured and packaged in a certified clean room.
  • Designed for aseptic fluid connections in bioprocessing and laboratory assemblies.
Catalog No. NC3440482 Supplier Accuflow Systems Supplier No. PFMS500PVDF
The Pure-Fit SIB Mini PVDF smooth inner-bore connector provides a seamless tubing-to-fitting transition for aseptic fluid transfer in bioprocessing and laboratory systems. Constructed from PVDF, the Mini design minimizes dead volume and reduces particle entrapment, supporting low extractables and biocompatibility.
  • Smooth inner bore reduces dead volume and particle entrapment.
  • PVDF construction for broad chemical compatibility and sterilization tolerance.
  • Mini series, 1/2 in (12.7 mm) size for common tubing applications.
  • Manufactured and packaged in a certified clean room.
  • Designed for aseptic fluid connections in bioprocessing and laboratory assemblies.