Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Tri-Clamp Fitting, PVDF, 5/8 in
Catalog No. NC3363117
Encompass_Preferred
Saint-Gobain Pure-Fit SIB Mini tri-clamp fitting in PVDF for hygienic fluid transfer. The Mini series fitting features a smooth inner bore and accepts tubing with a 5/8 in (15.9 mm) inner diameter, providing a sanitary transition for process lines.
  • Material: PVDF provides chemical resistance and low particle generation.
  • Fits 5/8 in (15.9 mm) inner-diameter tubing.
  • Mini tri-clamp configuration for sanitary connections.
  • Smooth inner bore reduces turbulence and particulate entrapment.
  • Suitable for hygienic and biopharmaceutical processing.
Catalog No. NC3363117 Supplier Accuflow Systems Supplier No. PFMS625PVDF
Saint-Gobain Pure-Fit SIB Mini tri-clamp fitting in PVDF for hygienic fluid transfer. The Mini series fitting features a smooth inner bore and accepts tubing with a 5/8 in (15.9 mm) inner diameter, providing a sanitary transition for process lines.
  • Material: PVDF provides chemical resistance and low particle generation.
  • Fits 5/8 in (15.9 mm) inner-diameter tubing.
  • Mini tri-clamp configuration for sanitary connections.
  • Smooth inner bore reduces turbulence and particulate entrapment.
  • Suitable for hygienic and biopharmaceutical processing.