Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich PFN-Br

Catalog No. 906980100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-698-0100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-698-0100MG Supplier Sigma Aldrich Supplier No. 906980100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PFN-Br is a solution processible, conjugated polymer electrolyte used as interficial layer in high performance organic photovoltaic cells (OPV) and organic light emitting diodes (OLED) to improve the interfacial properties.OPV devices with PFN-Br interfacial layer showed overall enhanced short-circuit current density, open-circuit voltage, fill factor and corresponding high efficiency in the inverted OPVs. This was attributed to the good contact between ZnO electron extraction layer and the active layer, good interface adhesion between the electron extraction layer and active layer, and enhanced charge transport via suppressed bimolecular recombination. Recent report of OPV with a record energy conversion efficiency of 17.3% also utilized PFN-Br, spin-coated on top of ZnO to improve the interfacial properties.Tandem Cell Device performance:ITO/ZnO/PFN-Br/PBDB-T:F-M/M-PEDOT/ZnO/PTB7- Th:O6T-4F:PC71BM/MoO3/Ag Voc=1.642 VJsc=14.35 mA/cm2FF=73.7% PCE=17.3% PFN-Br finds application in a wide range of areas:OPV interficial layer (metalic oxide compact layer)OLED electron transport layerPerovskite solar cellsFlexible printed electronicsCationic polymer electrolyteAnion exchange materialLight emitting electrochemical cellsRecommend to use 0.2 mg/ml concentration for electron-transport interfacial layer in OPV.

Specifications

CAS 889672-99-5
Linear Formula (C56H80N2.Br2)n
Quantity 100 mg
Synonym OS0995; PFN-P2; Poly(9,9-bis(3 -(N,N-dimethyl)-N-ethylammoinium-propyl-2,7-fluorene)-alt-2,7-(9,9-dioctylfluorene))dibromide
Molecular Weight (g/mol) 30,000-50,000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.