Learn More
AdipoGen PFOA ammonium salt
SDP
Catalog No. 502560174
Encompass_Preferred
Chemical. CAS 3825-26-1. Formula C8HF15O2 . NH3. Molecular Weight 431.1. Perfluorooctanoate PFOA is the dissociated carboxylate anion of perfluorooctanoic acid and its salts. PFOA is a stable, soluble surfactant primarily used in the form of the ammonium salt APFO as a processing aid in the production of various fluoropolymers. It is a very active fluorosurfactant with high stability.
Catalog No. 50-256-0174 Supplier AdipoGen Supplier No. CDXP0606G050
Chemical. CAS 3825-26-1. Formula C8HF15O2 . NH3. Molecular Weight 431.1. Perfluorooctanoate PFOA is the dissociated carboxylate anion of perfluorooctanoic acid and its salts. PFOA is a stable, soluble surfactant primarily used in the form of the ammonium salt APFO as a processing aid in the production of various fluoropolymers. It is a very active fluorosurfactant with high stability.